Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 6-Chloropyridine-2-carboxaldehyde, 95%, [54087-03-5], Z12581 141.555 C6H4ClNO

Catalog No. 5039398
Encompass_Preferred

Compound: 6-Chloropyridine-2-carboxaldehyde

CAS#: 54087-03-5

MDL#: not listed

Background: A chlorinated pyridine carbaldehyde used as a building block in pharmaceutical research. The chlorine substituent offers a reactive site for cross-coupling, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-393-98 Supplier Frontier Specialty Chemicals Supplier No. Z125811G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Chloropyridine-2-carboxaldehyde

CAS#: 54087-03-5

MDL#: not listed

Background: A chlorinated pyridine carbaldehyde used as a building block in pharmaceutical research. The chlorine substituent offers a reactive site for cross-coupling, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.