Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 6-Hydroxypyridazine-3-carboxylic acid, 95%, [37972-69-3], H12271 140.097 C5H4N2O3

Catalog No. 5037142
Encompass_Preferred

Compound: 6-Hydroxypyridazine-3-carboxylic acid

CAS#: 37972-69-3

MDL#: not listed

Background: A hydroxylated pyridazine carboxylic acid used as a building block in pharmaceutical research. The tautomeric hydroxyl/oxo group and the carboxylic acid group both allow for further heterocyclic derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-371-42 Supplier Frontier Specialty Chemicals Supplier No. H122711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Hydroxypyridazine-3-carboxylic acid

CAS#: 37972-69-3

MDL#: not listed

Background: A hydroxylated pyridazine carboxylic acid used as a building block in pharmaceutical research. The tautomeric hydroxyl/oxo group and the carboxylic acid group both allow for further heterocyclic derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.