Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, 7-Bromoquinoline, 0.92, [4965-36-0], B12115 208.055 C9H6BrN

Catalog No. 5035667
Encompass_Preferred

Compound: 7-Bromoquinoline

CAS#: 4965-36-0

MDL#: not listed

Background: A brominated quinoline used as a versatile building block in pharmaceutical and agrochemical synthesis. The bromine substituent provides a reactive site for palladium-catalyzed cross-coupling reactions to construct substituted quinoline scaffolds.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-356-67 Supplier Frontier Specialty Chemicals Supplier No. B121151G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 7-Bromoquinoline

CAS#: 4965-36-0

MDL#: not listed

Background: A brominated quinoline used as a versatile building block in pharmaceutical and agrochemical synthesis. The bromine substituent provides a reactive site for palladium-catalyzed cross-coupling reactions to construct substituted quinoline scaffolds.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.