Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Cyclohexylboronic acid pinacol ester, >95%, [87100-15-0], C1155 210.12 C12H23BO2

Catalog No. 50981824
Encompass_Preferred

Compound: Cyclohexylboronic acid pinacol ester

CAS#: 87100-15-0

MDL#: MFCD04038749

Background: Cyclohexylboronic acid pinacol ester is used as a ligand and coupling partner in Suzuki cross-coupling reactions in organic synthesis. Its pinacol ester form provides improved stability and handling compared to the free boronic acid, making it a useful research chemical for proteomics and synthetic chemistry applications.

Catalog No. 50-981-824 Supplier Frontier Specialty Chemicals Supplier No. C11551G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Cyclohexylboronic acid pinacol ester

CAS#: 87100-15-0

MDL#: MFCD04038749

Background: Cyclohexylboronic acid pinacol ester is used as a ligand and coupling partner in Suzuki cross-coupling reactions in organic synthesis. Its pinacol ester form provides improved stability and handling compared to the free boronic acid, making it a useful research chemical for proteomics and synthetic chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.