Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Cyclopentylboronic acid, 96%, [63076-51-7], C8488 113.95 C5H11BO2

Catalog No. 50982009
Encompass_Preferred

Compound: Cyclopentylboronic acid

CAS#: 63076-51-7

MDL#: MFCD01074541

Background: Cyclopentylboronic acid is a cycloalkylboronic acid used in palladium- and nickel-catalyzed cross-coupling reactions to construct functionalized aromatic and aliphatic hydrocarbons. It participates in reactions with aryl halides, tosylates, and mesylates, including Suzuki-type couplings used in pharmaceutical and fine chemical synthesis. It has also been applied in the arylation and alkylation of heterocyclic scaffolds such as diphenylisoxazole.

Catalog No. 50-982-009 Supplier Frontier Specialty Chemicals Supplier No. C84881G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Cyclopentylboronic acid

CAS#: 63076-51-7

MDL#: MFCD01074541

Background: Cyclopentylboronic acid is a cycloalkylboronic acid used in palladium- and nickel-catalyzed cross-coupling reactions to construct functionalized aromatic and aliphatic hydrocarbons. It participates in reactions with aryl halides, tosylates, and mesylates, including Suzuki-type couplings used in pharmaceutical and fine chemical synthesis. It has also been applied in the arylation and alkylation of heterocyclic scaffolds such as diphenylisoxazole.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.