Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Ethylboronic acid, 97%, [4433-63-0], E1104 73.89 C2H7BO2

Catalog No. 50982279
Encompass_Preferred

Compound: Ethylboronic acid

CAS#: 4433-63-0

MDL#: MFCD01074536

Background: Ethylboronic acid is used as an intermediate in the synthesis of bioactive molecules with anticancer, antiviral, and antibacterial properties, and its derivatives have been studied as inhibitors of bacterial beta-lactamases involved in antibiotic resistance. It is widely used as a reagent in Suzuki-Miyaura cross-coupling reactions, forming carbon-carbon bonds with aryl and vinyl halides under mild conditions, and is also used in analytical chemistry techniques such as chromatography and mass spectrometry.

Catalog No. 50-982-279 Supplier Frontier Specialty Chemicals Supplier No. E11041G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Ethylboronic acid

CAS#: 4433-63-0

MDL#: MFCD01074536

Background: Ethylboronic acid is used as an intermediate in the synthesis of bioactive molecules with anticancer, antiviral, and antibacterial properties, and its derivatives have been studied as inhibitors of bacterial beta-lactamases involved in antibiotic resistance. It is widely used as a reagent in Suzuki-Miyaura cross-coupling reactions, forming carbon-carbon bonds with aryl and vinyl halides under mild conditions, and is also used in analytical chemistry techniques such as chromatography and mass spectrometry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.