Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Mn(III) Protoporphyrin IX chloride, >95%, [120389-54-0], MNP562-9 651.03 C34H32ClMnN4O4

Catalog No. 5038507
Encompass_Preferred

Compound: Mn(III) Protoporphyrin IX chloride

CAS#: 120389-54-0

MDL#: MFCD00216788

Background: Mn(III) Protoporphyrin IX chloride is a metallated derivative of protoporphyrin IX that has been used as an experimental relaxation enhancement agent for MRI. Prostaglandin endoperoxide synthase reconstituted with Mn(III) protoporphyrin IX retains full cyclooxygenase activity, and manganese protoporphyrin IX-reconstituted myoglobin acts as an epoxidation catalyst with oxone. Electropolymerized manganese protoporphyrin IX has also been used to produce polyporphyrin films for supercapacitors.

Catalog No. 50-385-07 Supplier Frontier Specialty Chemicals Supplier No. MNP56291G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Mn(III) Protoporphyrin IX chloride

CAS#: 120389-54-0

MDL#: MFCD00216788

Background: Mn(III) Protoporphyrin IX chloride is a metallated derivative of protoporphyrin IX that has been used as an experimental relaxation enhancement agent for MRI. Prostaglandin endoperoxide synthase reconstituted with Mn(III) protoporphyrin IX retains full cyclooxygenase activity, and manganese protoporphyrin IX-reconstituted myoglobin acts as an epoxidation catalyst with oxone. Electropolymerized manganese protoporphyrin IX has also been used to produce polyporphyrin films for supercapacitors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.