Learn More
Frontier Specialty Chemicals Neopentylboronic acid, 701261-35-0, MFCD09750464, 1G
Supplier:  Frontier Specialty Chemicals N11521G
Encompass_Preferred
Empirical Formula (Hill Notation): C5H13BO2, Molecular Weight: 115.97, Synonym: 2,2-Dimethylpropylboronic acid, MP: 85-89 °C. Reactant for: Preparation of alkylarenes via tetraphosphine-palladium-catalyzed Suzuki cross-coupling with aryl halides.
Catalog No. 50-982-825