Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Pd(II) meso-Tetra(4-carboxyphenyl)porphine, 95%, [94288-44-5], PDT790 895.18 C48H28N4O8Pd

Catalog No. 5039033
Encompass_Preferred

Compound: Pd(II) meso-Tetra(4-carboxyphenyl)porphine

CAS#: 94288-44-5

MDL#: MFCD00672300

Background: Pd(II) meso-Tetra(4-carboxyphenyl)porphine is a synthetic porphyrin with unique photophysical properties useful for the detection of oxygen, carbon monoxide, and other analytes. It has been incorporated into hydrogels for lactate detection and into metal-organic frameworks for chemical sensing. Its carboxylic acid groups enable coupling to surfaces and biomolecules for sensor construction.

Catalog No. 50-390-33 Supplier Frontier Specialty Chemicals Supplier No. PDT7901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Pd(II) meso-Tetra(4-carboxyphenyl)porphine

CAS#: 94288-44-5

MDL#: MFCD00672300

Background: Pd(II) meso-Tetra(4-carboxyphenyl)porphine is a synthetic porphyrin with unique photophysical properties useful for the detection of oxygen, carbon monoxide, and other analytes. It has been incorporated into hydrogels for lactate detection and into metal-organic frameworks for chemical sensing. Its carboxylic acid groups enable coupling to surfaces and biomolecules for sensor construction.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.