Learn More
Worthington Biochemical Corporation 1G PEPSIN A
SDP
Catalog No. NC9675212
Encompass
Pepsin A,1 gm,Source:Porcine Stomach,Minimum Activity:=2500 units per mg dry weight,2X crystallized from dilute alcohol. A lyophilized powder. Store at 2-8degreeC,Unit Definition:1 unit releases 0.001 A280 as TCA soluble hydrolysis products
Catalog No. NC9675212 Supplier Worthington Biochemical Corporation Supplier No. LS003319
Pepsin A,1 gm,Source:Porcine Stomach,Minimum Activity:=2500 units per mg dry weight,2X crystallized from dilute alcohol. A lyophilized powder. Store at 2-8degreeC,Unit Definition:1 unit releases 0.001 A280 as TCA soluble hydrolysis products