Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Potassium methoxymethyltrifluoroborate, >95%, [910251-11-5], P10926 151.965 C2H5BF3KO

Catalog No. 5038742
Encompass_Preferred

Compound: Potassium methoxymethyltrifluoroborate

CAS#: 910251-11-5

MDL#: MFCD10566517

Background: A methoxymethyl-substituted organotrifluoroborate salt used as a coupling partner in Suzuki-type cross-coupling reactions. The ether linkage provides a stable, non-reactive substituent for pharmaceutical building block synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-387-42 Supplier Frontier Specialty Chemicals Supplier No. P109261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Potassium methoxymethyltrifluoroborate

CAS#: 910251-11-5

MDL#: MFCD10566517

Background: A methoxymethyl-substituted organotrifluoroborate salt used as a coupling partner in Suzuki-type cross-coupling reactions. The ether linkage provides a stable, non-reactive substituent for pharmaceutical building block synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.