Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Quinoxaline-6-boronic acid pinacol ester, 97%, [1167418-13-4], Q12133 256.108 C14H17BN2O2

Catalog No. 5039061
Encompass_Preferred

Compound: Quinoxaline-6-boronic acid pinacol ester

CAS#: 1167418-13-4

MDL#: MFCD11054040

Background: A quinoxaline boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It provides a convenient means to introduce a substituted quinoxaline scaffold into drug candidate synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-390-61 Supplier Frontier Specialty Chemicals Supplier No. Q121331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Quinoxaline-6-boronic acid pinacol ester

CAS#: 1167418-13-4

MDL#: MFCD11054040

Background: A quinoxaline boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It provides a convenient means to introduce a substituted quinoxaline scaffold into drug candidate synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.