Learn More
Med Vet International Rescue, Disinfectant Ready To Use, Liquid, One Step Disinfectant Cleaner And Deodorizer, 1 Gallon
Catalog No. 502827891
Encompass_Preferred
Quick 1 minute contact time against Canine Parvo, Panleukopenia, Ringworm, Kennel Cough and more. Safe to use around staff and animals. Compatible with a wide range of materials.
Catalog No. 50-282-7891 Supplier Med Vet International Supplier No. PHVRESCUELIQ
Add to Cart
Add to Cart
Quick 1 minute contact time against Canine Parvo, Panleukopenia, Ringworm, Kennel Cough and more. Safe to use around staff and animals. Compatible with a wide range of materials.