Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Vanadyl octaethylporphine, >95%, [27860-55-5], VOO534 599.7 C36H44N4OV

Catalog No. 5039388
Encompass_Preferred

Compound: Vanadyl octaethylporphine

CAS#: 27860-55-5

MDL#: MFCD00058868

Background: Vanadyl octaethylporphine is a synthetic porphyrin often used as a model compound for vanadyl petroporphyrins found in crude oil and geological samples. It has been studied for light-induced electron spin polarization and the characterization of excited quartet states, making it a useful reference compound in spectroscopy and photophysics research.

Catalog No. 50-393-88 Supplier Frontier Specialty Chemicals Supplier No. VOO5341G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Vanadyl octaethylporphine

CAS#: 27860-55-5

MDL#: MFCD00058868

Background: Vanadyl octaethylporphine is a synthetic porphyrin often used as a model compound for vanadyl petroporphyrins found in crude oil and geological samples. It has been studied for light-induced electron spin polarization and the characterization of excited quartet states, making it a useful reference compound in spectroscopy and photophysics research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.