Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1g, Zn(II) meso-Tetraphenylporphine (1-3% chlorin), >95%, [14074-80-7], T40942 678.129 C44H28N4Zn

Catalog No. 5039344
Encompass_Preferred

Compound: Zn(II) meso-Tetraphenylporphine (1-3% chlorin)

CAS#: 14074-80-7

MDL#: not listed

Background: A synthetic zinc porphyrin specialty chemical manufactured by Frontier Specialty Chemicals. It has been used as an ionophore for a cyanide ion-selective electrode and, together with quantum dots, to modify colloidal silica beads for colorimetric ammonia sensing.

Storage: Store at room temperature, protected from light.

Catalog No. 50-393-44 Supplier Frontier Specialty Chemicals Supplier No. T409421G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Zn(II) meso-Tetraphenylporphine (1-3% chlorin)

CAS#: 14074-80-7

MDL#: not listed

Background: A synthetic zinc porphyrin specialty chemical manufactured by Frontier Specialty Chemicals. It has been used as an ionophore for a cyanide ion-selective electrode and, together with quantum dots, to modify colloidal silica beads for colorimetric ammonia sensing.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.