Learn More
Labelmaster Inc 1-Gallon Closed Head Pail, Epoxy Phenolic Lining, 3/4" Spout
SDP
Catalog No. NC0997417
Encompass_Preferred
This Steel Closed Head Pail With Epoxy Phenolic Lining is a convenient way to ship liquid hazardous materials by cargo air, ground, rail and sea. The rugged and reinforced gray steel pail with 3/4" spout is virtually impenetrable and can protect your product with airtight seals and incredibly long life expectancies. The 24-gauge 1-gallon UN certified container can be used to ship Packing Group I, II, and III materials.
Catalog No. NC0997417 Supplier Labelmaster Inc Supplier No. KM1000EL
This Steel Closed Head Pail With Epoxy Phenolic Lining is a convenient way to ship liquid hazardous materials by cargo air, ground, rail and sea. The rugged and reinforced gray steel pail with 3/4" spout is virtually impenetrable and can protect your product with airtight seals and incredibly long life expectancies. The 24-gauge 1-gallon UN certified container can be used to ship Packing Group I, II, and III materials.