Learn More
US Plastic Corporation 1GAL JUG CLR PVC 38-400
SDP
Catalog No. NC0783679
Encompass
Clear PVC Jug. Medium-weight, industrial, clear jugs have a convenient handle with funnel top for easy pouring. Excellent clarity; Polypropylene cap with F217 liner; Corrosion resistant; 38/400 neck size; Lighter than glass jugs. Meets FDA standards. Cap is included.
Catalog No. NC0783679 Supplier US Plastic Corporation Supplier No. 66375
Clear PVC Jug. Medium-weight, industrial, clear jugs have a convenient handle with funnel top for easy pouring. Excellent clarity; Polypropylene cap with F217 liner; Corrosion resistant; 38/400 neck size; Lighter than glass jugs. Meets FDA standards. Cap is included.