Learn More
Avanti Polar Lipids 1GRM DOPC
Catalog No. NC9467752
Encompass
18:1 PC (cis), 1,2-Dioleoyl-sn-Glycero-3-Phosphocholine (DOPC); Powder; 1g; M.W. 705.93; CAS: 4235-95-4; Storage: Store in freezer at <-20 deg.C
Catalog No. NC9467752 Supplier Avanti Polar Lipids Supplier No. 850375P1G
18:1 PC (cis), 1,2-Dioleoyl-sn-Glycero-3-Phosphocholine (DOPC); Powder; 1g; M.W. 705.93; CAS: 4235-95-4; Storage: Store in freezer at <-20 deg.C