Learn More
Medchemexpress LLC 1H-1,2,3-triazole-4-carboxamide derivative (BIIB091) | 2247614-80-6 | 99.9% | C28H34N10O2 | 10MG
SDP
Catalog No. 5000258371
Encompass_Preferred
BIIB091 is a potent, selective, reversible Bruton's tyrosine kinase (BTK) inhibitor supplied as a high-purity small-molecule reagent for research and preclinical studies. It is used to probe BTK signaling and to evaluate therapeutic approaches in autoimmune and B-cell-mediated disease models. Not for human or clinical use.
  • Potent, selective reversible BTK inhibitor (IC50 <0.5 nM).
  • High purity (99.85% by HPLC).
  • Molecular formula C28H34N10O2; molecular weight 542.64.
  • Suitable for in vitro and preclinical assays.
  • Orally active profile reported in preclinical studies.
  • Supplied as a powder; soluble in DMSO.
Catalog No. 50-002-58371 Supplier Medchemexpress LLC Supplier No. HY13998410MG
BIIB091 is a potent, selective, reversible Bruton's tyrosine kinase (BTK) inhibitor supplied as a high-purity small-molecule reagent for research and preclinical studies. It is used to probe BTK signaling and to evaluate therapeutic approaches in autoimmune and B-cell-mediated disease models. Not for human or clinical use.
  • Potent, selective reversible BTK inhibitor (IC50 <0.5 nM).
  • High purity (99.85% by HPLC).
  • Molecular formula C28H34N10O2; molecular weight 542.64.
  • Suitable for in vitro and preclinical assays.
  • Orally active profile reported in preclinical studies.
  • Supplied as a powder; soluble in DMSO.