Learn More
Medchemexpress LLC 1h-1,2,3-Triazole, 5-(4-bromophenyl)- | 5301-98-4 | 99.6% | 250 MG
SDP
Catalog No. 5000341470
Encompass_Preferred
MetAP-2-IN-6 (4-(4-bromophenyl)-1H-1,2,3-triazole) is an inhibitor of MetAP2. It is intended for research related to conditions mediated by angiogenesis and is not validated for medical applications.
  • Inhibits MetAP2
  • Suitable for angiogenesis research
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • IC50 of 6 μM against HUVEC cell proliferation
  • IC50 of 20 μM against MS-1 cell proliferation
Catalog No. 50-003-41470 Supplier Medchemexpress LLC Supplier No. 250MGHY128147
MetAP-2-IN-6 (4-(4-bromophenyl)-1H-1,2,3-triazole) is an inhibitor of MetAP2. It is intended for research related to conditions mediated by angiogenesis and is not validated for medical applications.
  • Inhibits MetAP2
  • Suitable for angiogenesis research
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • IC50 of 6 μM against HUVEC cell proliferation
  • IC50 of 20 μM against MS-1 cell proliferation