Learn More
Medchemexpress LLC 1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one | 41443-28-1 | MFCD00792620 | >98.0% | 187.16 | C9H5N3O2 | 5 MG
SDP
Catalog No. 5000211708
Encompass_Preferred
ODQ (1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase (sGC) used as a research reagent in biochemical and cell-based studies. It blocks NO-activated sGC activity and has been reported to enhance the pro-apoptotic effects of cisplatin in certain cancer cell models. Supplied as a powder for preparation of stock solutions, it is intended for research use only.
  • Potent, selective inhibitor of soluble guanylyl cyclase.
  • Useful tool compound for studying nitric oxide-sGC signaling.
  • Reported to enhance cisplatin-induced apoptosis in some cell models.
  • Supplied as a powder suitable for preparing concentrated stock solutions.
  • Stable with defined storage recommendations for powder and solutions.
Catalog No. 50-002-11708 Supplier Medchemexpress LLC Supplier No. HY1012555MG
ODQ (1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase (sGC) used as a research reagent in biochemical and cell-based studies. It blocks NO-activated sGC activity and has been reported to enhance the pro-apoptotic effects of cisplatin in certain cancer cell models. Supplied as a powder for preparation of stock solutions, it is intended for research use only.
  • Potent, selective inhibitor of soluble guanylyl cyclase.
  • Useful tool compound for studying nitric oxide-sGC signaling.
  • Reported to enhance cisplatin-induced apoptosis in some cell models.
  • Supplied as a powder suitable for preparing concentrated stock solutions.
  • Stable with defined storage recommendations for powder and solutions.