Learn More
Medchemexpress LLC 1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one | 41443-28-1 | MFCD00792620 | 99.8% | 187.16 g/mol | C9H5N3O2 | 1 ML
SDP
Catalog No. 5000211707
Encompass_Preferred
ODQ (1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase (sGC) used in research to probe nitric oxide-cGMP signaling and related cellular processes. It is supplied in solution and solid formats for biochemical and cell-based studies.
  • Potent and selective inhibitor of soluble guanylyl cyclase (sGC).
  • Useful for studying nitric oxide-cGMP signaling in biochemical and cell-based assays.
  • Enhances pro-apoptotic effects of cisplatin in certain cell models.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes.
  • High purity (~99.8%) suitable for research applications.
Catalog No. 50-002-11707 Supplier Medchemexpress LLC Supplier No. HY10125510MM 1ML
ODQ (1H-[1,2,4]oxadiazolo[4,3-a]quinoxalin-1-one) is a potent, selective inhibitor of soluble guanylyl cyclase (sGC) used in research to probe nitric oxide-cGMP signaling and related cellular processes. It is supplied in solution and solid formats for biochemical and cell-based studies.
  • Potent and selective inhibitor of soluble guanylyl cyclase (sGC).
  • Useful for studying nitric oxide-cGMP signaling in biochemical and cell-based assays.
  • Enhances pro-apoptotic effects of cisplatin in certain cell models.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes.
  • High purity (~99.8%) suitable for research applications.