Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-1,2,4-Triazole-1-acetic acid, 97%

Catalog No. AAH31732MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31732MD 250 mg
AAH3173203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31732MD Supplier Thermo Scientific Chemicals Supplier No. H31732MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 28711-29-7 | C4H5N3O2 | 127.103 g/mol

1H-1,2,4-Triazole-1-acetic acid is used to produce (1-hydroxy-1-phosphono-2-[1,2,4]triazol-1-yl-ethyl)-phosphonic acid at the temperature of 100 - 110°C. It will need reagent H3PO4, PCl3 and solvent chlorobenzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-1,2,4-Triazole-1-acetic acid is used to produce (1-hydroxy-1-phosphono-2-[1,2,4]triazol-1-yl-ethyl)-phosphonic acid at the temperature of 100 - 110°C. It will need reagent H3PO4, PCl3 and solvent chlorobenzene

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28711-29-7
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00036051
InChI Key RXDBSQXFIWBJSR-UHFFFAOYSA-N
Synonym 1,2,4-triazole-1-acetic acid, 2-1h-1,2,4-triazol-1-yl acetic acid, 1h-1,2,4-triazole-1-acetic acid, 1,2,4 triazol-1-yl-acetic acid, unii-w3ask2wbnh, 1,2,4-triazol-1-ylacetic acid, w3ask2wbnh, 1h-1,2,4-triazol-1-ylacetic acid, 1h-1,2,4-triazol-1-yl acetic acid, 1h-1,2,4-triazole-1-aceticacid
PubChem CID 1810180
IUPAC Name 2-(1,2,4-triazol-1-yl)acetic acid
SMILES C1=NN(C=N1)CC(=O)O

Specifications

Melting Point 201°C to 205°C
Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 127.1
Percent Purity 97%
Chemical Name or Material 1H-1,2,4-Triazole-1-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.