Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-1,2,4-triazole-1-ethanol, β-[(4-chlorophenyl)methyl]-α-methyl-α-phenyl-, (αR,βS)- | 259200-30-1 | 99.8% | 10 MG
SDP

Catalog No. 5000233546
Encompass_Preferred

(2R,3S)-Brassinazole is the enantiomeric form of brassinazole, a triazole-type inhibitor of brassinosteroid biosynthesis used in plant biology research. It is supplied as a high-purity solid and as a ready-to-use DMSO solution for in vitro and in vivo studies.

  • High purity: 99.8% with 100% enantiomeric excess.
  • Chemical formula C18H18ClN3O; molecular weight 327.81 g/mol.
  • Available as solid (5 mg, 10 mg) and as 10 mM in DMSO solution.
  • Highly soluble in DMSO (150 mg/mL); in vivo formulation clear at ≥ 3.75 mg/mL.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-33546 Supplier Medchemexpress LLC Supplier No. HY121161C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2R,3S)-Brassinazole is the enantiomeric form of brassinazole, a triazole-type inhibitor of brassinosteroid biosynthesis used in plant biology research. It is supplied as a high-purity solid and as a ready-to-use DMSO solution for in vitro and in vivo studies.

  • High purity: 99.8% with 100% enantiomeric excess.
  • Chemical formula C18H18ClN3O; molecular weight 327.81 g/mol.
  • Available as solid (5 mg, 10 mg) and as 10 mM in DMSO solution.
  • Highly soluble in DMSO (150 mg/mL); in vivo formulation clear at ≥ 3.75 mg/mL.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.