Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-1,2,4-triazole-3-carboxylic acid | 4928-87-4 | MFCD00078268 | 97.0% | 113.07 g·mol⁻¹ | C3H3N3O2 | 5 G
SDP

Catalog No. 5000244501
Encompass_Preferred

1H-1,2,4-triazole-3-carboxylic acid (CAS 4928-87-4) is a small-molecule biochemical reagent supplied at 97% purity for research use. It is used as a building block in organic synthesis and as a reagent in biochemical assays. Molecular formula C3H3N3O2; molecular weight 113.07 g·mol⁻¹.

  • Purity 97% for research and assay use.
  • Carboxylic acid functional group useful in derivatization and coupling reactions.
  • Small-molecule profile suitable for synthetic and analytical workflows.
  • Molecular weight 113.07 g·mol⁻¹ and formula C3H3N3O2 for easy reference.
  • Available in 5 G packaging for laboratory-scale experiments.

Catalog No. 50-002-44501 Supplier Medchemexpress LLC Supplier No. HY1078705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1H-1,2,4-triazole-3-carboxylic acid (CAS 4928-87-4) is a small-molecule biochemical reagent supplied at 97% purity for research use. It is used as a building block in organic synthesis and as a reagent in biochemical assays. Molecular formula C3H3N3O2; molecular weight 113.07 g·mol⁻¹.

  • Purity 97% for research and assay use.
  • Carboxylic acid functional group useful in derivatization and coupling reactions.
  • Small-molecule profile suitable for synthetic and analytical workflows.
  • Molecular weight 113.07 g·mol⁻¹ and formula C3H3N3O2 for easy reference.
  • Available in 5 G packaging for laboratory-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.