Learn More
Medchemexpress LLC 1H-1,4,7-triazonine-1,4,7-triacetic acid, α1-(2-carboxyethyl)hexahydro-, tris(t-Bu) ester | 1190101-34-8 | MFCD29083820 | 99.8% | 543.69 g/mol | C27H49N3O8 | 100 MG
SDP
Catalog No. 5000283893
Encompass_Preferred
NODAGA-tris(t-Bu ester) is a tri-tert-butyl ester-protected NODAGA chelator intended for conjugation to biomolecules and subsequent radiolabeling for positron emission tomography (PET) studies. It provides a protected, bifunctional scaffold that facilitates attachment to peptides, proteins, and antibodies and enables stable chelation of radiometals after deprotection.
  • Protected NODAGA chelator for conjugation to biomolecules.
  • Suitable for labeling peptides, proteins, and antibodies.
  • Designed for radiolabeling workflows for PET imaging.
  • High purity (99.8%) with RP-HPLC documentation available.
  • Available as solid and as DMSO solutions for convenience.
  • Molecular formula C27H49N3O8; molecular weight 543.69 g/mol.
Catalog No. 50-002-83893 Supplier Medchemexpress LLC Supplier No. HYW539893100MG
NODAGA-tris(t-Bu ester) is a tri-tert-butyl ester-protected NODAGA chelator intended for conjugation to biomolecules and subsequent radiolabeling for positron emission tomography (PET) studies. It provides a protected, bifunctional scaffold that facilitates attachment to peptides, proteins, and antibodies and enables stable chelation of radiometals after deprotection.
  • Protected NODAGA chelator for conjugation to biomolecules.
  • Suitable for labeling peptides, proteins, and antibodies.
  • Designed for radiolabeling workflows for PET imaging.
  • High purity (99.8%) with RP-HPLC documentation available.
  • Available as solid and as DMSO solutions for convenience.
  • Molecular formula C27H49N3O8; molecular weight 543.69 g/mol.