Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-1,4,7-triazonine-1,4,7-triacetic acid, α1-(2-carboxyethyl)hexahydro-, tris(tert-butyl) ester | 1190101-34-8 | MFCD29083820 | 99.8% | 543.69 g/mol | C27H49N3O8 | 50 MG
SDP

Catalog No. 5000283898
Encompass_Preferred

NODAGA-tris(t-Bu ester) is a tert-butyl-protected bifunctional chelator precursor used for covalent attachment to biomolecules. It is designed to conjugate to peptides, proteins, or antibodies; after deprotection the chelator can be radiolabeled for PET imaging studies. The reagent is supplied as a high-purity research chemical suitable for labeling workflows and analytical characterization.

  • Tert-butyl protected chelator precursor suitable for conjugation to peptides and antibodies.
  • Compatible with deprotection and subsequent radiolabeling for PET imaging.
  • High purity as measured by HPLC, appropriate for research use.
  • Soluble in DMSO for formulation and conjugation workflows.
  • Molecular weight 543.69 g/mol; chemical formula C27H49N3O8.

Catalog No. 50-002-83898 Supplier Medchemexpress LLC Supplier No. HYW53989350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

NODAGA-tris(t-Bu ester) is a tert-butyl-protected bifunctional chelator precursor used for covalent attachment to biomolecules. It is designed to conjugate to peptides, proteins, or antibodies; after deprotection the chelator can be radiolabeled for PET imaging studies. The reagent is supplied as a high-purity research chemical suitable for labeling workflows and analytical characterization.

  • Tert-butyl protected chelator precursor suitable for conjugation to peptides and antibodies.
  • Compatible with deprotection and subsequent radiolabeling for PET imaging.
  • High purity as measured by HPLC, appropriate for research use.
  • Soluble in DMSO for formulation and conjugation workflows.
  • Molecular weight 543.69 g/mol; chemical formula C27H49N3O8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.