Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,11H-Eicosafluoro-1-undecanol 90.0+%, TCI America™
SDP

Catalog No. E023910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E023910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E023910G Supplier TCI America Supplier No. E023910G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 307-70-0
Molecular Formula C11H4F20O
Molecular Weight (g/mol) 532.12
MDL Number MFCD00039628
InChI Key GJYLSFCFHFUODO-UHFFFAOYSA-N
Synonym 1h,1h,11h-eicosafluoro-1-undecanol, 1-undecanol, 1h,1h,11h-eicosafluoro, 1,1,11-trihydroperfluoroundecanol, 1h,1h,11h-perfluoro-1-undecanol, 1h,1h,11h-eicosafluoroundecanol-1, 1h,1h,11h-eicosafluoro-1-undecanol, tech., 1h,11h-eicosafluoro-1-undecanol, 1h,1h,11h-perfluoroundecan-1-ol, 1-undecanol,1h,11h-eicosafluoro, 1h, 1h, 11h-eicosafluoroundecanol
PubChem CID 67548
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol
SMILES C(C(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Color White
Quantity 10 g
Formula Weight 532.12
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H,11H-Eicosafluoro-1-undecanol
EINECSNumber (2)-0287
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.