Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Nonafluorohexyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. N09775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. N09775G Supplier TCI America Supplier No. N09775G

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 52591-27-2
Molecular Formula C9H7F9O2
Molecular Weight (g/mol) 318.14
MDL Number MFCD00236104
InChI Key GYUPEJSTJSFVRR-UHFFFAOYSA-N
Synonym Acrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Acrylate, Acrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
PubChem CID 104247
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C

Specifications

Color Colorless
UN Number 3272
Quantity 5 g
Formula Weight 318.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluorohexyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-4056
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.