Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™

Catalog No. AAL1658914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658914 25 g
AAL1658906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1658914 Supplier Thermo Scientific Chemicals Supplier No. L1658914

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-86-1
Molecular Formula C12H5F21O
Molecular Weight (g/mol) 564.14
MDL Number MFCD00039545
InChI Key FLXYIZWPNQYPIT-UHFFFAOYSA-N
Synonym 2-perfluorodecyl ethanol, 1h,1h,2h,2h-perfluorododecan-1-ol, 1,1,2,2-tetrahydroperfluoro dodecanol, 1h,1h,2h,2h-perfluoro-1-dodecanol, unii-a72u7q973t, 1h,1h,2h,2h-perfluorododecanol, 1,1,2,2-tetrahydroperfluoro-1-dodecanol, 1h,1h,2h,2h-perfluorodeca-1-ol, 1h,1h,2h,2h-perfluoro-1-dodecanol 1g
PubChem CID 70083
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
SMILES OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 90°C to 95°C
Boiling Point 145°C (10 mmHg)
Linear Formula CF3(CF2)9CH2CH2OH
Quantity 25 g
Beilstein 2029867
Solubility Information Insoluble in water.
Formula Weight 564.14
Percent Purity 96%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
EINECSNumber 212-748-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.