Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H,2H-Perfluoro-1-Octanol

Catalog No. 37053325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-053-325G 25 g
37-053-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 37-053-325G Supplier Sigma Aldrich Supplier No. 37053325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H,2H,2H-Perfluoro-1-octanol is a fluorotelomer alcohol.1H,1H,2H,2H-Perfluoro-1-octanol may be used to generate droplets in droplet-based microfluidics systems. It can be used as a starting material in the synthesis of 1H,1H,2H,2H -perfluorooctyl vinyl ether (FOVE).

Specifications

CAS 647-42-7
Density 1.651 g/mL at 25°C
Boiling Point 88°C to 95°C28 mmHg, lit.)
Molecular Formula C8H5F13O
Refractive Index n20/D 1.313
Linear Formula CF3(CF2)5CH2CH2OH
MDL Number MFCD00042143
Quantity 25 g
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol; 6:2 FTOH; 1H,1H,2H,2H-Perfluoro-1-octanol
Molecular Weight (g/mol) 364.1 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.