Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H,2H-Perfluorodecyl acrylate

Catalog No. 4744875ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-448-75ML 5 mL
47-448-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-448-75ML Supplier Sigma Aldrich Supplier No. 4744875ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H,2H,2H-Perfluorodecyl acrylate

Specifications

CAS 27905-45-9
Density 1.637 g/mL (at 25°C)
Boiling Point 90°C (4 mmHg)
Molecular Formula C13H7F17O2
Linear Formula H2C=CHCO2CH2CH2(CF2)7 CF3
Refractive Index n20/D 1.337 (literature)
MDL Number MFCD00042306
Quantity 5 mL
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl acrylate; 1 H,1 H,2 H,2 H-Perfluorodecyl acrylate
Molecular Weight (g/mol) 518.17
Percent Purity 97%
Biological Activity Liver
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.