Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Perfluorodecyltrichlorosilane, 96%, Thermo Scientific™

Catalog No. AAL1658403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1658403 1 g
AAL1658406 5 g
AAL1658414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AAL1658403 Supplier Thermo Scientific Chemicals Supplier No. L1658403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluorodecyltrichlorosilane is a halogenated compound used for biochemical research. It is applied to movable microparts of microelectromechanical systems , thereby reducing the surface energy and preventing the stickiness. Further, it is used to coat micro- and nano-features on stamps for a nanoimprint lithography. In addition, it is used in the preparation of (1H,1H,2H,2H-heptadecafluorodecyl)trimethoxysilane by reacting with methanol.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Moisture sensitive. Incompatible with oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78560-44-8
Molecular Formula C10H4Cl3F17Si
Molecular Weight (g/mol) 581.55
MDL Number MFCD00042344
InChI Key VIFIHLXNOOCGLJ-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorodecyltrichlorosilane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl silane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl, 1h,1h,2h,2h-perfluorotrichlorosilane, trichloro 1h,1h,2h,2h-perfluorodecyl silane, trichloro 1h,1h,2h,2h-heptadecafluorodecyl silane, 1-trichlorosilyl-1h,1h,2h,2h-heptadecafluorodecane, 1h,1h,2h,2h-perfluorodecyltrichlorosilane 1g, heptadecafluoro-1,1,2,2-tetrahydrodecyl trichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-trichlorosilane
PubChem CID 123577
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 224°C
Physical Form Liquid
Quantity 1 g
Flash Point >200°C (392°F)
Refractive Index 1.348
Sensitivity Moisture sensitive
Density 1.54
Linear Formula CF3(CF2)7(CH2)2SiCl3
UN Number UN2987
Beilstein 5889653
Solubility Information Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents.
Formula Weight 581.56
Percent Purity 96%
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyltrichlorosilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.