Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H,2H-Perfluorodecyltriethoxysilane

Catalog No. 6587585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-875-85G 5 g
65-875-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-875-85G Supplier Sigma Aldrich Supplier No. 6587585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H,2H,2H-Perfluorodecyltriethoxysilane(PFDTES) is a fluoroalkylsilanes(FAS) with low surface energy and high wettability due to the presence of fluorinated carbons. It is used as a superhydrophobic agent for synthesizing various functional coatings.PFDTES is used for the superhydrophobicity of polyvinylidene fluoride(PVDF) to protect the metal surfaces from fouling. A cost effective and scalable superamphiphobic spray coating of PFDTES on stainless steel surfaces may potentially be used to create anti-icing and stain-resistant surfaces. It may be used to modify the surface of poly(methyl methacrylate(PMMA) for the fabrication of microfluidic chips.

Specifications

CAS 101947-16-4
Density 1.389 g/mL (at 25°C)
Boiling Point 209°C to 230°C
Molecular Formula C16H19F17O3Si
Refractive Index n20/D 1.342
Linear Formula C16H19F17O3Si
Quantity 5 g
MDL Number MFCD00042334
Synonym Triethoxy-1 H,1 H,2 H,2 H-perfluorodecylsilane
Molecular Weight (g/mol) 610.38
Percent Purity 97%
Biological Activity Liver
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.