Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H,2H-Perfluorododecyltrichlorosilane

Catalog No. 7299655G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-996-55G 5 g
72-996-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-996-55G Supplier Sigma Aldrich Supplier No. 7299655G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H,2H,2H-Perfluorododecyltrichlorosilane

Specifications

CAS 102488-49-3
Melting Point 50°C to 55°C
Molecular Formula C12H4Cl3F21Si
Linear Formula CF3(CF2)9 CH2CH2SiCl3
Quantity 5 g
MDL Number MFCD07368770
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluorododecyltrichlorosilane
Molecular Weight (g/mol) 681.57
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.