Learn More
Strem, An Ascensus Company CAS# 2043-47-2. 10g. 1H,1H,2H,2H-Perfluorohexan-1-ol, 99%. MFCD00039543
SDP
Catalog No. 5090109397
Encompass
CAS# 2043-47-2. 10g. 1H,1H,2H,2H-Perfluorohexan-1-ol, 99%. MFCD00039543. Molecular Weight: 264.09. Molecular Formula: F(CF2)4CH2CH2OH. Color/form: colorless liq. Strem# 09-6085. http://www.strem.com/catalog/v/09-6085/
Catalog No. 50-901-09397 Supplier Strem, An Ascensus Company Supplier No. 09608510G
CAS# 2043-47-2. 10g. 1H,1H,2H,2H-Perfluorohexan-1-ol, 99%. MFCD00039543. Molecular Weight: 264.09. Molecular Formula: F(CF2)4CH2CH2OH. Color/form: colorless liq. Strem# 09-6085. http://www.strem.com/catalog/v/09-6085/