Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Perfluorooctanol, 97%

Catalog No. AAB2015618
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2015618 50 g
AAB2015609 10 g
AAB2015630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2015618 Supplier Thermo Scientific Chemicals Supplier No. B2015618
  • $484.50
    $411.65 / Each of 1

    Save $72.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 647-42-7 | C8H5F13O | 364.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctanol is used to prepare fluorinated aromatic diamine by reacting with 2-(4-nitrophenoxy)-4-nitrophenol through Mitsunobu reaction, which one reacts with dianhydrides such as 4,4′-(hexafluoroisopropylidene)diphthalic anhydride (6FDA), pyromellitic dianhydride (PMDA) and benzophenonetetracarboxylic dianhydride (BTDA) to prepare polyimides.

Solubility
Miscible with chloroform and methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 647-42-7
Molecular Formula C8H5F13O
Molecular Weight (g/mol) 364.106
MDL Number MFCD00042143
InChI Key GRJRKPMIRMSBNK-UHFFFAOYSA-N
Synonym 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol, 2-perfluorohexyl ethanol, 1h,1h,2h,2h-perfluorooctan-1-ol, 1h,1h,2h,2h-perfluorooctanol, 1h,1h,2h,2h-perfluoro-1-octanol, unii-g2r5yo5n3v, 1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, 1h,1h,2h,2h-tridecafluoro-1-n-octanol, g2r5yo5n3v, 1,1,2,2-tetrahydroperfluoro-1-octanol
PubChem CID 69537
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctan-1-ol
SMILES C(CO)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.68
Boiling Point 90°C (20 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.314
Quantity 50 g
Beilstein 1992757
Solubility Information Miscible with chloroform and methanol. Immiscible with water.
Formula Weight 364.11
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctanol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-477-1
RTECSNumber RH1460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.