Alfa Aesar™ 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, 97%

Manufacturer: Alfa Aesar™ 4454304

Quantity 2g

 View more versions of this product

Catalog No. AA4454304

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102488-47-1
    Molecular Formula C10H10ClF13Si
    Formula Weight 440.71
    MDL Number MFCD00042359
    UN Number UN2986
    Beilstein 7040835
    Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane

    Specifications

    Formula Weight 440.71
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Physical Form Liquid
    Molecular Formula C10H10ClF13Si
    MDL Number MFCD00042359
    CAS 102488-47-1
    Beilstein 7040835
    Quantity 2g
    Refractive Index 1.3453
    Sensitivity Moisture Sensitive
    UN Number UN2986
    Boiling Point 189° to 191°C
    Color Colorless
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Density 1.461g/mL
    Flash Point 52°C (180°F)
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
    Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
    Assay 97%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit