Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H,2H-Perfluorooctyltriethoxysilane

Catalog No. 66742025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-742-025G 25 g
66-742-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 66-742-025G Supplier Sigma Aldrich Supplier No. 66742025G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethoxy(1H,1H,2H,2H-perfluoro-1-octyl)silane (POTS) is a fluorine based polymer. POTS possesses low surface free energy leading to its anti-adhesive behavior to polar and non-polar substances. It shows very good weathering stability on account of the carbon- fluorine bond. POTS hydrolyzes in wet environment to form silane based film and self healing anticorrosive polymers, the film formed from the hydrolysis and polycondensation of POTS and water is hydrophobic in nature. This wetting property acts in repelling aqueous electrolyte solution away from metallic substrates and hence providing corrosion protection to metal substrate.POTS can be used as a silane based coating on poly(vinylidene fluoride) (PVDF) to synthesize a membrane for the removal of hazardous volatile organic compounds. Glass substrates can be modified with POTS to increase the contact angle for 3D reactive inkjet printing of polydimethylsiloxane (PDMS). It can also be used in the synthesis of superhydrophobic carbon nanotubes(CNT) hollow membranes for membrane desalination (MD) as eco-friendly products of fresh water. It is deposited on the roughened surface of titanium to render it superhydrophobic. POTS may be used as a component of fluorine modified ORMOCER (Organically Modified Ceramics) coating materials.

Specifications

CAS 51851-37-7
Density 1.3299 g/mL (at 25°C)
Molecular Formula C14H19F13O3Si
Refractive Index n20/D 1.344
Linear Formula C14H19F13O3Si
Quantity 25 g
MDL Number MFCD00042333
Synonym Triethoxy(1 H,1 H,2 H,2 H-perfluoro-1-octyl)silane; Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octyl)silane
Molecular Weight (g/mol) 510.36
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.