Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Tridecafluoro-n-octyl Acrylate (stabilized with HQ + MEHQ) 98.0+%, TCI America™
SDP

Catalog No. T34515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34515G 5 g
T345125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34515G Supplier TCI America Supplier No. T34515G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17527-29-6
Molecular Formula C11H7F13O2
Molecular Weight (g/mol) 418.15
MDL Number MFCD00042351
InChI Key VPKQPPJQTZJZDB-UHFFFAOYSA-N
Synonym Acrylic Acid 1H,1H,2H,2H-Tridecafluoro-n-octyl Ester
PubChem CID 87149
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCOC(=O)C=C

Specifications

Color Colorless
Boiling Point 50°C
Quantity 5 g
Formula Weight 418.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Tridecafluoro-n-octyl Acrylate (stabilized with HQ + MEHQ)
EINECSNumber (2)-4064
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.