Learn More
Cayman Chemical 1H 1H 2H 2HPRfluoroctnsu 500mg
SDP
Catalog No. 502915089
Encompass_Preferred
A polyfluoroalkyl substance; has been found as a contaminant in sea and river water
Catalog No. 50-291-5089 Supplier Cayman Chemical Supplier No. 37260500 MG
A polyfluoroalkyl substance; has been found as a contaminant in sea and river water