Learn More
Cayman Chemical 1H,1H,2H,2H-Perfluorodecanesulfonic Acid, Purity: ≥80%, CAS: 39108-34-4, 50 mg
SDP
Catalog No. 502919452
Encompass_Preferred
A polyfluoroalkyl substance; has been found as a contaminant in sea and river water
Catalog No. 50-291-9452 Supplier Cayman Chemical Supplier No. 3725950 MG
A polyfluoroalkyl substance; has been found as a contaminant in sea and river water