Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H,2H-Perfluoro-1-Hexene

Catalog No. 37145925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-145-925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 37-145-925G Supplier Sigma Aldrich Supplier No. 37145925G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H,2H-Perfluoro-1-Hexene

Specifications

CAS 13814-75-0
Molecular Formula Cl4MoO
Refractive Index n20/D 1.3
Linear Formula MoOCl4
Quantity 5 g
MDL Number MFCD00014222
Synonym Molybdenum chloride oxide; Molybdenum oxide chloride; Molybdenum oxytetrachloride; Molybdenum tetrachloride monoxide; Molybdenum(VI) oxychloride; Tetrachlorooxomolybdenum
Molecular Weight (g/mol) 253.75 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.