Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. M143325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M143325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M143325G Supplier TCI America Supplier No. M143325G

missing translation for 'validMainframeMsg'

Specifications

CAS 355-93-1
Color Yellow
Boiling Point 88°C
Molecular Formula C9H8F8O2
MDL Number MFCD00039278
Quantity 25 g
Synonym 1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
InChI Key ZNJXRXXJPIFFAO-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 300.15
PubChem CID 67739
Formula Weight 300.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with TBC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.