Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,8H,8H-Dodecafluoro-1,8-octanediol, 97%, Thermo Scientific™

Catalog No. AAL1648403
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1648403 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1648403 Supplier Thermo Scientific Chemicals Supplier No. L1648403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,8H,8H-Dodecafluoro-1,8-octanediol is used in pharma industry.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 90177-96-1
Boiling Point 105°C (0.08mmHg)
Molecular Formula C8H6F12O2
Linear Formula HOCH2(CF2)6CH2OH
Quantity 1g
MDL Number MFCD00153145
Synonym 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-perfluorooctane-1,8-diol, 1,8-dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane, 1,8-octanediol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro, 1h,1h,8h,8h-perfluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluoro-1,8-octanediol, 1h,1h,8h,8h-dodecafluorooctane-1,8-diol, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octane-1,8-diol
Solubility Information Soluble in methanol.
InChI Key XZJPYETUABEQFI-UHFFFAOYSA-N
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
Molecular Weight (g/mol) 362.12
PubChem CID 3820533
Formula Weight 362.12
Percent Purity 97%
Chemical Name or Material 1H,1H,8H,8H-Dodecafluoro-1,8-octanediol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.