Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,9H-Perfluoro-1-nonanol, 97%, Thermo Scientific Chemicals

Catalog No. AAB2140722
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2140722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2140722 Supplier Thermo Scientific Chemicals Supplier No. B2140722
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-18-1
Molecular Formula C9H4F16O
Molecular Weight (g/mol) 432.104
MDL Number MFCD00039629
InChI Key MSXVQELLSMPBFD-UHFFFAOYSA-N
Synonym 1h,1h,9h-hexadecafluoro-1-nonanol, hexadecafluoro-1-nonanol, 1-nonanol, hexadecafluoro, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 1-nonanol, 1h,1h,9h-hexadecafluoro, 1h,1h,9h-hexadecafluoro-1-hydroxynonane, 1h,1h,9h-perfluoro-1-nonanol, 1h,1h,9h-hexadecafluorononan-1-ol, omega-h-hexadekafluornonanol-1 german, 1h,1h,9h-hexadecafluorononanol
PubChem CID 9779
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol
SMILES C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 55°C to 60°C
Boiling Point 157°C to 158°C (200 mmHg)
Quantity 100 g
Beilstein 1808149
Formula Weight 432.11
Percent Purity 97%
Chemical Name or Material 1H,1H,9H-Perfluoro-1-nonanol
EINECSNumber 206-806-0
RTECSNumber RA7905000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.