Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H-Heptadecafluoro-1-nonanol 97.0+%, TCI America™
SDP

Catalog No. H12325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12325G Supplier TCI America Supplier No. H12325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 423-56-3
Molecular Formula C9H3F17O
Molecular Weight (g/mol) 450.10
MDL Number MFCD00153183
InChI Key BSXJTDJJVULBTQ-UHFFFAOYSA-N
Synonym 1h,1h-perfluoro-1-nonanol, 1h,1h-perfluorononanol, 1h,1h-heptadecafluoro-1-nonanol, 1h,1h-perfluorononan-1-ol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-1-nonanol, heptadecafluorooctylmethanol, 1h,1h-heptadecafluorononanol, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro, 1-nonanol,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluoro-1-nonanol #
PubChem CID 533991
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononan-1-ol
SMILES OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 69°C
Color White
Boiling Point 176°C
Quantity 5 g
Formula Weight 450.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1H,1H-Heptadecafluoro-1-nonanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.