Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H,1H-Perfluorooctyl methacrylate

Catalog No. 7764915G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-649-15G 5 g
77-649-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-649-15G Supplier Sigma Aldrich Supplier No. 7764915G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H,1H-Perfluorooctyl methacrylate

Specifications

CAS 3934-23-4
Density 1.574 g/mL (at 25°C)
Molecular Formula C12H7F15O2
Linear Formula C12H7F15O2
MDL Number MFCD00039247
Quantity 5 g
Synonym 1,1-Dihydroperfluorooctyl methacrylate; 1 H,1 H-Pentadecafluorooctyl methacrylate; 1 H,H-Pentadecafluoro-1-octyl methacrylate; 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octyl methacrylate; Methacrylic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester; Nyebar
Molecular Weight (g/mol) 468.16
Percent Purity 96%
Biological Activity Liver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.