Learn More
Medchemexpress LLC 1H-5,10b-Propeno-1,7-phenanthrolin-8(7H)-one, 2,3,4,4a,5,6-hexahydro-12-methyl-... | 103548-82-9 | 256.34 | 5 MG
SDP
Catalog No. 5000415124
Encompass_Preferred
Huperzine B is a Lycopodium alkaloid isolated from *Huperzia serrata* and acts as a highly selective acetylcholinesterase (AChE) inhibitor. It can be used to improve Alzheimer's disease.
  • Isolated from *Huperzia serrata*.
  • Highly selective acetylcholinesterase (AChE) inhibitor.
  • Can be used to improve Alzheimer's disease.
  • For research use only.
  • Purity 98.73%.
Catalog No. 50-004-15124 Supplier Medchemexpress LLC Supplier No. HYN20435MG
Huperzine B is a Lycopodium alkaloid isolated from *Huperzia serrata* and acts as a highly selective acetylcholinesterase (AChE) inhibitor. It can be used to improve Alzheimer's disease.
  • Isolated from *Huperzia serrata*.
  • Highly selective acetylcholinesterase (AChE) inhibitor.
  • Can be used to improve Alzheimer's disease.
  • For research use only.
  • Purity 98.73%.