Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-5-Mercapto-1,2,3-triazole, Sodium Saltdihydrate 97%, Thermo Scientific™

Catalog No. AC133560250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133560250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC133560250 Supplier Thermo Scientific Chemicals Supplier No. 133560250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59032-27-8
Molecular Formula C2H2N3NaS
Molecular Weight (g/mol) 123.109
InChI Key HHGMADGROXARPN-UHFFFAOYSA-M
Synonym sodium 1,2,3-triazole-5-thiolate, 5-mercapto-1,2,3-triazole monosodium salt, sodium 1h-1,2,3-triazol-5-ylsulfanide, 1h-1,2,3-triazole-5-thiol sodium salt, 1h-5-mercapto-1,2,3-triazole,sodium salt, tas:1h-1,2,3-triazole-4-thiol sodium salt, sodium 1h-1,2,3-triazole-4-thiolate, sodium 1h-1,2,3-triazole-5-thiolate, sodium 2h-1,2,3-triazole-4-thiolate, 1h-1,2,3-triazole-4-thiol sodium salt
PubChem CID 23685427
IUPAC Name sodium;2H-triazole-4-thiolate
SMILES C1=NNN=C1[S-].[Na+]

Specifications

Color Green
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 159.14
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1H-5-Mercapto-1,2,3-triazole, sodium saltdihydrate, 97%
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.